Learn More
Tricaprin, TRC
Specifications
Specifications
| Chemical Name or Material | Tricaprin |
| CAS | 621-71-6 |
| Molecular Formula | C33 H62 O6 |
| Quantity | 10 g |
| Synonym | Decanoic acid 1,1',1''-(1,2,3-propanetriyl) ester, tri-Caprin, Decanoic acid 1,2,3-propanetriyl ester, tri-Decanoin, Actor M 2, Capric acid triglyceride, Capric triglyceride, Captex 1000, Decanoic acid triglyceride, Dynasan 110, Glycerin tricaprate, Glycerin tridecanoate, Glycerol tricaprate, Glycerol tricaprin, Glycerol tridecanoate, Glyceryl tricaprate, NSC 147475, Panacet 1000, Tri(decanoyl)glycerol, Tricapric glyceride, Tricaprin, Tridecanoin, Triglyceride DDD |
| InChI Formula | InChI=1S/C33H62O6/c1-4-7-10-13-16-19-22-25-31(34)37-28-30(39-33(36)27-24-21-18-15-12-9-6-3)29-38-32(35)26-23-20-17-14-11-8-5-2/h30H,4-29H2,1-3H3 |
| SMILES | CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC |
| IUPAC Name | 2,3-di(decanoyloxy)propyl decanoate |
| Molecular Weight (g/mol) | 554.84 |
| Formula Weight | 554.4546 |