Learn More
Tricaprin 8000 μg/mL in Pyridine, Dr. Ehrenstorfer
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | Tricaprin |
| CAS | 621-71-6 |
| Analyte or Component Names | Tricaprin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H62 O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Decanoic acid 1,1',1''-(1,2,3-propanetriyl) ester, tri-Caprin, Decanoic acid 1,2,3-propanetriyl ester, tri-Decanoin, Actor M 2, Capric acid triglyceride, Capric triglyceride, Captex 1000, Decanoic acid triglyceride, Dynasan 110, Glycerin tricaprate, Glycerin tridecanoate, Glycerol tricaprate, Glycerol tricaprin, Glycerol tridecanoate, Glyceryl tricaprate, NSC 147475, Panacet 1000, Tri(decanoyl)glycerol, Tricapric glyceride, Tricaprin, Tridecanoin, Triglyceride DDD |
| InChI Formula | InChI=1S/C33H62O6/c1-4-7-10-13-16-19-22-25-31(34)37-28-30(39-33(36)27-24-21-18-15-12-9-6-3)29-38-32(35)26-23-20-17-14-11-8-5-2/h30H,4-29H2,1-3H3 |
| SMILES | CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC |
| IUPAC Name | 2,3-di(decanoyloxy)propyl decanoate |
| Show More |