Learn More
BRAND™ Triangular Wedge-Shape PTFE Magnetic Stirring Bars
Product Code. 10762351 Shop All Brand Products
Click to view available options
Length (Metric):
25 mm
35 mm
40 mm
50 mm
55 mm
80 mm
Diameter (Metric):
8 mm
10 mm
12 mm
14 mm
16 mm
Unit Size:
Pack of 10
Product Code. 10762351
Brand: BRAND™ 137812
Only null left
Add to basket
Add to basket
An excellent option for liquid agitation in the laboratory.
  • Ideal for dissolving solids and mixing of sediments due to scraping action
  • Angular surface creates high turbulence even at low speeds
  • Provided in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Description Triangular wedge-shaped
Diameter (Metric) 14 mm
Length (Metric) 25 mm
Material PTFE
Quantity 10/Pk.
Shape Triangle