Learn More
Triamcinolone acetonide, MedChemExpress
Product Code. p-200096751
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16472249
Brand: Medchem Express HYB0636
MedChemExpress Triamcinolone acetonide is a more potent type of triamcinolone, being about 8 times as effective as prednisone.
Triamcinolone acetonide is a more potent type of triamcinolone, being about 8 times as effective as prednisone.
Specifications
Chemical Name or Material Triamcinolone acetonide
CAS 76-25-5
Color White
Purity Grade Notes Research
Molecular Formula C24H31FO6
Quantity 1 g
Solubility Information DMSO : 50 mg/mL (115.07 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.23 mM; Need ultrasonic)
SMILES OCC([C@]12[C@@]3([C@@]([C@@]4([H])[C@]([C@H](C3)O)([C@@]5(C(CC4)=CC(C=C5)=O)C)F)([H])C[C@@]1([H])OC(C)(O2)C)C)=O
Molecular Weight (g/mol) 434.5
Formula Weight 434.5
Percent Purity 99.4%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.