Learn More
tri-Sodium Phosphate Dodecahydrate, Supelco™
Specifications
Specifications
| Melting Point | 75°C (Elimination of Water of Crystallisation) |
| CAS | 10101-89-0 |
| Quantity | 1 kg |
| MDL Number | MFCD00149198 |
| Synonym | tri-Sodium phosphate dodecahydrate, Sodium phosphate tribasic, Trisodium phosphate, tri-Sodium phosphate dodecahydrate |
| Molecular Weight (g/mol) | 380.12 |
| Grade | Research |
| Density | 1.62 g/cm3 at 20°C |
| Chemical Name or Material | Tri-Sodium Phosphate Dodecahydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Informativa sulla privacy.
Individuate un'opportunità di miglioramento?