Learn More
tri-Sodium citrate dihydrate, BAKER ANALYZED™, J.T. Baker™, Avantor™
Shop All J.T.Baker Products
Click to view available options
:
500 g
Specifications
Specifications
| pH | 7.0 to 9.0 (5% Solution at 25°C) |
| Quantity | 500 g |
| Assay Percent Range | min. 99.0% |