Learn More
Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent
Product Code. 11537851 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 1 mL ampules
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11537851 10 x 1 mL ampules Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11537851 Supplier Thermo Scientific™ Supplier No. TS49016
 Restricted Item
Estimated Shipment: 26-10-2026
Employ a catalyzed silylation reagent formulation, for derivatization of hydroxyls with the Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent.
Employ a catalyzed silylation reagent formulation, for derivatization of hydroxyls with the Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent. Composed of TMSI/BSA/TMCS (3:3:2), this catalyzed silylation reagent formulation typically reacts with hydroxyls in a short period of time at 60–80 °C to form TMS ethers. Highly hindered hydroxyls may require several hours.
  • Converts all classes of hydroxyl groups to TMS ethers
  • Under usual conditions, the reaction is complete in a short period of time at 60 to 80°C, although very hindered hydroxyls may require several hours
TRUSTED_SUSTAINABILITY
Specifications
For Use With GC
Hazards Toxic
Standard Concentration 25 - 45, 25 - 45, 20 - 25%
Storage Room Temperature
Quantity 10 x 1 mL ampules
Standard Components Trimethylsilyl N-trimethylsilylacetamidate, 1-(Trimethylsilyl)-1 H-imidazole, Chlorotrimethylsilane
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-Sil TBT, 10 x 1 mL
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use to extinguish.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Trimethylchlorosilane, N-(trimethylsilyl)imidazole