Learn More
Tri(4-phenylbutyryl)glycerol, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Tri(4-phenylbutyryl)glycerol |
| CAS | 611168-24-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C33H38O6 |
| Quantity | 50 mg |
| Synonym | Benzenebutanoic Acid 1,1',1''-(1,2,3-Propanetriyl) Ester, Benzenebutanoic Acid 1,2,3-Propanetriyl Ester, Glycerol Phenylbutyrate, HPN 100 |
| InChI Formula | InChI=1S/C33H38O6/c34-31(22-10-19-27-13-4-1-5-14-27)37-25-30(39-33(36)24-12-21-29-17-8-3-9-18-29)26-38-32(35)23-11-20-28-15-6-2-7-16-28/h1-9,13-18,30H,10-12,19-26H2 |
| SMILES | O=C(OCC(OC(CCCC1=CC=CC=C1)=O)COC(CCCC2=CC=CC=C2)=O)CCCC3=CC=CC=C3 |
| IUPAC Name | 2,3-bis(4-phenylbutanoyloxy)propyl 4-phenylbutanoate |
| Molecular Weight (g/mol) | 530.65 |
| Show More |