Learn More
Invitrogen™ TRF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581990
Description
Immunogen sequence: STGKEDKQPA PGPVEKPPRE PARQLRNPPT TIGMMTLKAA FKTLSGAQDS EAAFAKLDQK DLVLPTQALP ASPALKNKRP RKDENESSAP ADGEGGSELQ PKNKRMTISR LVLEEDSQST EPS Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 74%.
TRF2 is a ubiquitously expressed protein that is implicated in the control of telomere length. TRF2, like TRF1 contains a Myb-related DNA binding motif. It binds to duplex TTAGGG repeats and is localized to all human telomeres in metaphase chromosomes. TRF2 is thought to protect chromosome ends by maintaining the correct structure at telomere termini. The use of mutant forms of TRF2 has implicated a role for TRF2 in the prevention of senescence in primary human cells. Recently, it has been shown that inhibition of TRF2 resulted in apoptosis in a subset of mammalian cell types.
Specifications
| TRF2 | |
| Polyclonal | |
| Unconjugated | |
| TERF2 | |
| Telomeric DNA-binding protein; telomeric repeat binding factor 2; telomeric repeat binding protein 2; telomeric repeat-binding factor 2; TERF2; TRBF2; TRF2; TTAGGG repeat-binding factor 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7014 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15554 | |
| TERF2 | |
| Recombinant protein corresponding to Human TERF2. Recombinant protein control fragment (Product #RP-101881). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.