Learn More
Uvex Trend 95188 Insole - Width 11
Comfortable climatic insole with antistatic properties for Uvex 2 Trend
Brand:  Uvex 9518851
Additional Details : Weight : 0.11000kg
 View more versions of this product
Product Code. 17277981
  • 222.00 NOK / 1 pair
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Full surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No. 9518.8
  • Size: 38 to 52
Specifications
Specifications
Black
Microvelour
No
Wide width
No
Uvex 2 Trend
51
Unisex
Yes