Learn More
Trenbolone Enanthate, TRC
Product Code. 30452861
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452861 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30452861 Supplier Toronto Research Chemicals Supplier No. TRCT719030100MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Este artículo no se puede devolver. Vea la política de devoluciones
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trenbolone enanthate
CAS 1629618-98-9
Molecular Formula C25 H34 O3
Quantity 100 mg
Synonym (17β)-17-[(1-Oxoheptyl)oxy]-estra-4,9,11-trien-3-one, Trenbolone Enanthate
InChI Formula InChI=1S/C25H34O3/c1-3-4-5-6-7-24(27)28-23-13-12-22-21-10-8-17-16-18(26)9-11-19(17)20(21)14-15-25(22,23)2/h14-16,21-23H,3-13H2,1-2H3/t21-,22+,23+,25+/m1/s1
SMILES CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CCC4=C3C=C[C@]12C
IUPAC Name [(8S,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl] heptanoate
Molecular Weight (g/mol) 382.54
Formula Weight 382.2508