Learn More
TREM1 Antibody (L5-B8.2A12.3A12) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-40907JF635
This item is not returnable.
View return policy
Description
TREM1 Monoclonal antibody specifically detects TREM1 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Frozen),CyTOF,Immunocytochemistry/ Immunofluorescence
Specifications
| TREM1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| Mouse TREM-1 Fc fusion protein | |
| 0.25 mL | |
| Signal Transduction | |
| 54210 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), CyTOF, Immunocytochemistry/Immunofluorescence | |
| L5-B8.2A12.3A12 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |