missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM1 Antibody (L5-B8.2A12.3A12) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-40907-0.05MG
This item is not returnable.
View return policy
Description
TREM1 Monoclonal specifically detects TREM1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| TREM1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready | |
| Q9JKE2 | |
| TREM1 | |
| Mouse TREM-1 Fc fusion protein. | |
| 0.05 mg | |
| Signal Transduction | |
| 54210 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
| CyTOF, Immunofluorescence, Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry | |
| L5-B8.2A12.3A12 | |
| Unconjugated | |
| Phosphate buffered saline with No Preservative | |
| CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction