missing translation for 'onlineSavingsMsg'
Learn More

TRBV5-4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13524-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16891774

  • 3468.15 NOK / 150µL
Estimated Shipment: 15-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TRBV5-4
Polyclonal
Unconjugated
TRBV5-4
TCRBV5S4, TCRBV5S6A3N2T, TRBV5 4, TRBV54, TRBV5-4
Rabbit
Antigen Affinity Chromatography
RUO
28611
-20°C
Liquid
Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A0A0C4DH59
TRBV5-4
TRBV5-4 Fusion Protein Ag4337
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers