Learn More
IKA™ Trayster Basics
Product Code. 13417669 Shop All IKA Products
Change view
Click to view available options
Speed Range:
0 to 20 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range unitSize
13417669 0 to 20 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13417669 Supplier IKA™ Supplier No. 0004005000
Only null left
Add to basket
Add to basket
Trayster Basic Overhead shaker with vertical rotation for smooth but effective mixing with a fixed speed.
  • Ideal for biological samples, particularly blood samples.
  • Suitable for separating and mixing processes of powdered and liquid samples in Eppendorf tubes and Greiner tubes up to 50 mL.
  • The easy removable attachments allow simple and quick cleaning of the unit of spilled samples.
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 0 to 20 rpm
Electrical Requirements 100/240 V, 24 VDC, 50/60Hz, 1A
Voltage 100/240 V
Temperature Range (Metric) 4°C to 50°C
Amperage 1 A
Certifications/Compliance IP 21 DIN EN 60529
Frequency 50/60 Hz
Depth (Metric) 140 mm
Width (Metric) 460 mm
Model Trayster Basic
Humidity Range 80%