Learn More
Ohaus™ Tray Pad Set
Product Code. 15772659 Shop All Ohaus Products
Click to view available options
Color:
Black
Unit Size:
Each
Product Code. 15772659
Brand: Ohaus™ 30400245
Only null left
Add to basket
Add to basket
OHAUS™ Tray Pad Set offers added padding to multi-tube vortex mixers.
  • OHAUS offers a large selection of accessories that can be used on all OHAUS vortex mixers
  • Increase throughput by mixing multiple samples at one time
  • Effortlessly mix up to 52 microtubes or eight microplates
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tray Pad
Color Black
Description Tray Pad Set - Top And Bottom