Learn More
Buerkle™ Transport Trolley
Product Code. 10346131 Shop All Bürkle Products
Change view
Click to view available options
For Use With (Equipment):
600 x 400 mm Containers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10346131 600 x 400 mm Containers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10346131 Supplier Buerkle™ Supplier No. 34149900
  • ONLINE EXCLUSIVE PRICE
    1580.00 NOK
    1431.12 NOK / Pack of 1
    Save 148.88 NOK
    9% Off
Estimated Shipment: 21-12-2026
For use with Buerkle™ stacking/storage containers
  • Flat platform trolley with swivel castors for high manoeuvrability
  • High load capacity with low dead weight
TRUSTED_SUSTAINABILITY
Specifications
Product Type Transport Trolley
Color Red
Description Equipment for storage and stacking containers
For Use With (Equipment) 600 x 400 mm Containers
Length (English) 23.62 in.
Length (Metric) 600 mm
Width (English) 15.75 in.
Width (Metric) 400 mm