Learn More
Fisherbrand™ Transport Box XL Tube Inserts
GREENER_CHOICE
Product Code. 17916260 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
10 mL Blood Tubes and 15 mL Conical Tubes
3.5 to 10 mL Blood Tubes and 15/50 mL Conical Tubes
3.5 to 7.5 mL Blood Tubes
Unit Size:
Pack of 3
Product Code. Holds unitSize
17916260 10 mL Blood Tubes and 15 mL Conical Tubes Pack of 3
17926260 3.5 to 7.5 mL Blood Tubes Pack of 3
17956260 3.5 to 10 mL Blood Tubes and 15/50 mL Conical Tubes Pack of 3
3 options
Product Code. 17916260
Transport Box XL Tube Rack Insert Accessories
  • Designed to fit Fisherbrand Transport Box XL
  • Thermoformed PETG
  • Tube Rack Inserts can be used separately
  • Lid removes completely for easy cleaning
  • Insert deep enough to be filled with ice
Specifications
Product Type Rack Insert
Color Clear
Material PETG
Dimensions (L x W x H) 13.1 x 14.3 x 5.6 in.
Holds 10 mL Blood Tubes and 15 mL Conical Tubes
Quantity 3 Pk.