Description
All dimensions are approximate.
- Transparent, colorless
- Terylene mesh reinforced wall
- Flexible yet great inherent strength
- Suitable for moderate pressure or vacuum applications
Specifications
Specifications
| Description | 8 x 3mm |
| Material | PVC |
| Overall Length (Metric) | 10 m |
| For Use With (Application) | Suitable For Moderate Pressure Or Vacuum Applications |
| Packaging Quantity | 1 Pack |
| Color | Transparent |