Learn More
Vitlab™ Transparent Polypropylene Funnel
Click to view available options
Capacity (Metric):
14 mL
32 mL
88 mL
222 mL
342 mL
817 mL
Unit Size:
Each
Pack of 12
Pack of 24
Description
- Transparent polypropylene funnel
- Rapid flow due to a steep 60° angle
Specifications
Specifications
| Diameter (Metric)Top | 40mm |
| Capacity (Metric) | 14 mL |
| Diameter (Metric) Stem | 4mm |
| Material | Polypropylene |
| Height (Metric) | 65 mm |
| Angle | 60° |
| Autoclavable | Autoclavable |
| For Use With (Application) | Suitable for food contact |
| Length (Metric) Stem | 35mm |