Learn More
Vitlab™ Transparent Polypropylene Funnel
Product Code. 10622811
Change view
Click to view available options
Capacity (Metric):
14 mL
32 mL
88 mL
222 mL
342 mL
817 mL
Unit Size:
Each
Pack of 12
Pack of 24
Product Code. Capacity (Metric) unitSize
10622811 222 mL Each
10708171 14 mL Each
11827960 14 mL Pack of 24
12724098 222 mL Pack of 12
10465811 32 mL Each
11817960 32 mL Pack of 24
10632811 342 mL Each
10608972 342 mL Pack of 12
10058540 817 mL Each
10783923 817 mL Pack of 12
10242261 88 mL Each
11 options
Product Code. 10622811
Brand: Vitlab™ 41494
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these funnels for a variety of bottling and decanting applications in the food industry. VITLAB™ Transparent Polypropylene Funnels feature a practical handle with loop for hanging. Ideal for powders and liquids.
  • Transparent polypropylene funnel
  • Rapid flow due to a steep 60° angle
Specifications
Diameter (Metric)Top 100mm
Capacity (Metric) 222 mL
Diameter (Metric) Stem 8mm
Material Polypropylene
Height (Metric) 155 mm
Angle 60°
Autoclavable Autoclavable
For Use With (Application) Suitable for food contact
Length (Metric) Stem 77mm