Learn More
Vitlab™ Transparent Polypropylene Funnel
Description
- Transparent polypropylene funnel
- Rapid flow due to a steep 60° angle
Specifications
Specifications
| Diameter (Metric)Top | 150mm |
| Capacity (Metric) | 817 mL |
| Diameter (Metric) Stem | 15mm |
| Material | Polypropylene |
| Height (Metric) | 220 mm |
| Angle | 60° |
| Autoclavable | Autoclavable |
| For Use With (Application) | Suitable for food contact |
| Length (Metric) Stem | 95mm |