Learn More
Vitlab™ Transparent Polypropylene Beaker
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 12
Pack of 6
Description
- Highly transparent polypropylene beaker
- To preserve markings, do not clean at temperatures exceeding 60°C
- Conditionally autoclavable at 121°C (2 bar) according to DIN EN 285
- For autoclaving, we recommend model 440941 with raised graduations
Specifications
Specifications
| Diameter (Metric) Outer | 70 mm |
| Capacity (Metric) | 250 mL |
| Material | Polypropylene |
| Height (Metric) | 120 mm |