Learn More
Fisherbrand™ Translucent Platinum-Cured Silicone Tubing
Product Code. 11778148 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.5 mm
0.8 mm
1.6 mm
2.4 mm
3.2 mm
4.8 mm
6.4 mm
8 mm
12.8 mm
16 mm
Diameter (Metric) Outer:
2.1 mm
2.4 mm
3.2 mm
4 mm
4.8 mm
6.4 mm
8 mm
9.6 mm
12.8 mm
16 mm
22.4 mm
Overall Length (Metric):
5 m
25 m
Unit Size:
25m
5m
Each
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Overall Length (Metric) unitSize
11778148 8 mm 12.8 mm 5 m 5m
11998199 1.6 mm 3.2 mm 5 m 5m
11988199 0.5 mm 2.1 mm 5 m 5m
11718178 16 mm 22.4 mm 5 m Each
11768088 0.8 mm 2.4 mm 25 m Each
11788088 1.6 mm 3.2 mm 25 m Each
11758088 0.8 mm 2.4 mm 5 m Each
11778108 2.4 mm 4 mm 25 m Each
11788108 3.2 mm 4.8 mm 5 m Each
11718118 3.2 mm 6.4 mm 25 m Each
11768128 4.8 mm 8 mm 5 m Each
11708118 3.2 mm 6.4 mm 5 m Each
11778128 4.8 mm 8 mm 25 m 25m
11768108 2.4 mm 4 mm 5 m Each
11748118 4.8 mm 6.4 mm 5 m Each
11708138 6.4 mm 9.6 mm 5 m Each
11718138 6.4 mm 9.6 mm 25 m 25m
11758168 12.8 mm 16 mm 5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Product Code. 11778148
  • ONLINE EXCLUSIVE PRICE
    288.00 NOK
    222.74 NOK / 5m
    Save 65.26 NOK
    22% Off
View all promotions
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Use this tubing for pharmaceutical, biotech, peristaltic pumping and food applications. Fisherbrand™ Translucent Platinum-Cured Silicone Tubing is odorless, nontoxic and features excellent heat resistance.
  • Translucent
  • Non-yellowing
  • No peroxide by-products
  • Standard high tear strength
  • High gloss finish
  • Excellent quality
  • Highly hydrophobic
  • Extended life in Peristaltic pumps
Specifications
Certifications/Compliance FDA, BFR and WRAS approved
Autoclavable Autoclavable
Diameter (Metric) Inner 8 mm
Material Platinum-cured Silicone
Diameter (Metric) Outer 12.8 mm
Overall Length (Metric) 5 m
Thickness (Metric) 2.4 mm
For Use With (Application) Biological , pharmaceutical, biotechnology, aerospace, hot work
Min. Operating Temperature (Metric) -40°C
Sterility Sterile
Color Translucent
Flexibility Flexible
Max. Operating Temperature (Metric) 200°C