Learn More
Fisherbrand™ Translucent Platinum-Cured Silicone Tubing
Product Code. 11768128 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.5 mm
0.8 mm
1.6 mm
2.4 mm
3.2 mm
4.8 mm
6.4 mm
8 mm
12.8 mm
16 mm
Diameter (Metric) Outer:
2.1 mm
2.4 mm
3.2 mm
4 mm
4.8 mm
6.4 mm
8 mm
9.6 mm
12.8 mm
16 mm
22.4 mm
Overall Length (Metric):
25 m
5 m
Unit Size:
25m
5m
Each
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Overall Length (Metric) unitSize
11768128 4.8 mm 8 mm 5 m Each
11998199 1.6 mm 3.2 mm 5 m 5m
11988199 0.5 mm 2.1 mm 5 m 5m
11718178 16 mm 22.4 mm 5 m Each
11768088 0.8 mm 2.4 mm 25 m Each
11788088 1.6 mm 3.2 mm 25 m Each
11758088 0.8 mm 2.4 mm 5 m Each
11778108 2.4 mm 4 mm 25 m Each
11788108 3.2 mm 4.8 mm 5 m Each
11718118 3.2 mm 6.4 mm 25 m Each
11708118 3.2 mm 6.4 mm 5 m Each
11778128 4.8 mm 8 mm 25 m 25m
11768108 2.4 mm 4 mm 5 m Each
11748118 4.8 mm 6.4 mm 5 m Each
11708138 6.4 mm 9.6 mm 5 m Each
11778148 8 mm 12.8 mm 5 m 5m
11718138 6.4 mm 9.6 mm 25 m 25m
11758168 12.8 mm 16 mm 5 m Each
18 options
Product Code. 11768128
Only null left
Add to basket
Add to basket
Use this tubing for pharmaceutical, biotech, peristaltic pumping and food applications. Fisherbrand™ Translucent Platinum-Cured Silicone Tubing is odorless, nontoxic and features excellent heat resistance.
  • Translucent
  • Non-yellowing
  • No peroxide by-products
  • Standard high tear strength
  • High gloss finish
  • Excellent quality
  • Highly hydrophobic
  • Extended life in Peristaltic pumps
Specifications
Certifications/Compliance FDA, BFR and WRAS approved
Autoclavable Autoclavable
Diameter (Metric) Inner 4.8 mm
Material Platinum-cured Silicone
Diameter (Metric) Outer 8 mm
Overall Length (Metric) 5 m
Thickness (Metric) 1.6 mm
For Use With (Application) Biological , pharmaceutical, biotechnology, aerospace, hot work
Min. Operating Temperature (Metric) -40°C
Sterility Sterile
Color Translucent
Flexibility Flexible
Max. Operating Temperature (Metric) 200°C