Learn More
BRAND™ Translucent Polypropylene Y-shape Tubing Connector
Product Code. 12110062 Shop All Brand Products
Change view
Click to view available options
Tubing Size:
10 to 11 mm tubing I.D.
3 mm tubing I.D.
4 to 5 mm tubing I.D.
6 to 7 mm tubing I.D.
8 to 9 mm tubing I.D.
Unit Size:
Pack of 20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size unitSize
12110062 4 to 5 mm tubing I.D. Pack of 20
15271509 6 to 7 mm tubing I.D. Pack of 20
11833930 3 mm tubing I.D. Pack of 20
11843970 8 to 9 mm tubing I.D. Pack of 20
10579461 10 to 11 mm tubing I.D. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12110062 Supplier BRAND™ Supplier No. 152903
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Constructed of translucent polypropylene. Accommodates three tubes with Y-shaped stem and arms.
  • Translucent polypropylene construction
  • Accommodates 3 tubes of the same diameter
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Overall Length (Metric) 35 mm
Tubing Size 4 to 5 mm tubing I.D.
Width (Metric) 21 mm
Shape Y
Color Translucent
Product Type Tubing Connector