Learn More
- Home
- Products
- Tubing
- Rubber and Silicone Tubing
- Fisherbrand™ Translucent Platinum Cured Silicone Tubing
Fisherbrand™ Translucent Platinum Cured Silicone Tubing
Product Code. p-8272091 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.5 mm
3.2 mm
4 mm
4.8 mm
6 mm
6.4 mm
8 mm
9.5 mm
9.6 mm
12.8 mm
16 mm
16.0 mm
19.2 mm
Diameter (Metric) Outer:
2.1 mm
4.8 mm
5.6 mm
6.4 mm
8 mm
11.2 mm
11.9 mm
12.8 mm
14.4 mm
15.9 mm
16 mm
17.6 mm
20.8 mm
22.4 mm
25.4 mm
Maximum Pressure:
0.5 bar
1 bar
1.5 bar
1bar
2 bar
30 bar
35 bar
40 bar
Overall Length (Metric):
25 m
5 m
Unit Size:
25m
Each
| Product Code. | Diameter (Metric) Inner | Diameter (Metric) Outer | Maximum Pressure | Overall Length (Metric) | unitSize |
|---|---|---|---|---|---|
| 11728138 | 6.4 mm | 11.2 mm | 1bar | 5 m | Each |
| 11748088 | 0.5 mm | 2.1 mm | 2 bar | 25 m | Each |
| 11788168 | 12.8 mm | 17.6 mm | 0.5 bar | 25 m | Each |
| 11708178 | 16 mm | 20.8 mm | 0.5 bar | 25 m | Each |
| 11798108 | 3.2 mm | 4.8 mm | 0.5 bar | 25 m | Each |
| 11758118 | 4.8 mm | 6.4 mm | 0.5 bar | 25 m | Each |
| 11738118 | 4 mm | 5.6 mm | 0.5 bar | 25 m | Each |
| 11798128 | 6.4 mm | 8 mm | 0.5 bar | 25 m | Each |
| 11763974 | 6 mm | 11.9 mm | 40 bar | 25 m | Each |
| 11768148 | 8 mm | 11.2 mm | 0.5 bar | 25 m | Each |
| 11788148 | 8 mm | 12.8 mm | 1 bar | 25 m | Each |
| 11708158 | 8 mm | 14.4 mm | 1.5 bar | 25 m | Each |
| 11773974 | 8 mm | 14.4 mm | 35 bar | 25 m | Each |
| 11783974 | 9.5 mm | 15.9 mm | 30 bar | 25 m | Each |
| 11728158 | 9.6 mm | 12.8 mm | 0.5 bar | 25 m | Each |
| 11748158 | 9.6 mm | 14.4 mm | 1 bar | 25 m | 25m |
| 11748168 | 9.6 mm | 16 mm | 1 bar | 25 m | Each |
| 11758138 | 6.4 mm | 12.8 mm | 1 bar | 25 m | Each |
| 11728178 | 16.0 mm | 22.4 mm | 1 bar | 25 m | Each |
| 11708188 | 19.2 mm | 25.4 mm | 0.5 bar | 25 m | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Autoclavable
- Non-yellowing
- High gloss finish
- Excellent quality
- Highly hydrophobic
- No peroxide byproducts
- High tear strength as standard
- Extended life in Peristaltic pumps
- Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Specifications
Specifications
| Certifications/Compliance | FDA, BFR and WRAS approved |
| Diameter (Metric) Inner | 6.4 mm |
| Diameter (Metric) Outer | 11.2 mm |
| Overall Length (Metric) | 5 m |
| Maximum Pressure | 1bar |