Learn More
Fisherbrand™ Translucent Platinum Cured Silicone Tubing
Product Code. 11728138 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.5 mm
3.2 mm
4 mm
4.8 mm
6 mm
6.4 mm
8 mm
9.5 mm
9.6 mm
12.8 mm
16 mm
16.0 mm
19.2 mm
Diameter (Metric) Outer:
2.1 mm
4.8 mm
5.6 mm
6.4 mm
8 mm
11.2 mm
11.9 mm
12.8 mm
14.4 mm
15.9 mm
16 mm
17.6 mm
20.8 mm
22.4 mm
25.4 mm
Maximum Pressure:
0.5 bar
1 bar
1.5 bar
1bar
2 bar
30 bar
35 bar
40 bar
Overall Length (Metric):
5 m
25 m
Unit Size:
25m
Each
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Maximum Pressure Overall Length (Metric) unitSize
11728138 6.4 mm 11.2 mm 1bar 5 m Each
11748088 0.5 mm 2.1 mm 2 bar 25 m Each
11788168 12.8 mm 17.6 mm 0.5 bar 25 m Each
11708178 16 mm 20.8 mm 0.5 bar 25 m Each
11798108 3.2 mm 4.8 mm 0.5 bar 25 m Each
11758118 4.8 mm 6.4 mm 0.5 bar 25 m Each
11738118 4 mm 5.6 mm 0.5 bar 25 m Each
11798128 6.4 mm 8 mm 0.5 bar 25 m Each
11763974 6 mm 11.9 mm 40 bar 25 m Each
11768148 8 mm 11.2 mm 0.5 bar 25 m Each
11788148 8 mm 12.8 mm 1 bar 25 m Each
11708158 8 mm 14.4 mm 1.5 bar 25 m Each
11773974 8 mm 14.4 mm 35 bar 25 m Each
11783974 9.5 mm 15.9 mm 30 bar 25 m Each
11728158 9.6 mm 12.8 mm 0.5 bar 25 m Each
11748158 9.6 mm 14.4 mm 1 bar 25 m 25m
11748168 9.6 mm 16 mm 1 bar 25 m Each
11758138 6.4 mm 12.8 mm 1 bar 25 m Each
11728178 16.0 mm 22.4 mm 1 bar 25 m Each
11708188 19.2 mm 25.4 mm 0.5 bar 25 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 11728138
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Providing a number of advantages over conventional peroxide cured silicones. Fisherbrand™ Translucent Platinum Cured Silicone Tubing is available in a comprehensive range of stock sizes.
  • Autoclavable
  • Non-yellowing
  • High gloss finish
  • Excellent quality
  • Highly hydrophobic
  • No peroxide byproducts
  • High tear strength as standard
  • Extended life in Peristaltic pumps
  • Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Specifications
Certifications/Compliance FDA, BFR and WRAS approved
Diameter (Metric) Inner 6.4 mm
Diameter (Metric) Outer 11.2 mm
Overall Length (Metric) 5 m
Maximum Pressure 1bar