Learn More
Translucent Crucibles
Product Code. 12897733
Change view
Click to view available options
Diameter (Metric) Outer:
29mm
38mm
43mm
57mm
67mm
82mm
Unit Size:
Each
Product Code. Diameter (Metric) Outer unitSize
12897733 43mm Each
12857723 29mm Each
12867723 38mm Each
12897723 57mm Each
12807733 67mm Each
12817733 82mm Each
6 options
Product Code. 12897733
Brand: Yorlab 10446/11
Only null left
Specifications
Color Translucent
Material Silica
Capacity (Metric) 30 mL
Diameter (Metric) Outer 43mm
Height (Metric) 39mm
Product Type Crucible