Learn More
Yorlab™ Translucent Crucibles
Product Code. 12867723
Change view
Click to view available options
Diameter (Metric) Outer:
29mm
38mm
43mm
57mm
67mm
82mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
12867723 38mm Each
12857723 29mm Each
12897733 43mm Each
12897723 57mm Each
12807733 67mm Each
12817733 82mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12867723 Supplier Yorlab™ Supplier No. 10444/16
  • ONLINE EXCLUSIVE PRICE
    348.00 NOK
    284.35 NOK / Pack of 1
    Save 63.65 NOK
    18% Off
Estimated Shipment: 22-12-2026
Specifications
Color Translucent
Material Silica
Capacity (Metric) 10 mL
Diameter (Metric) Outer 38mm
Height (Metric) 19mm
Product Type Crucible