missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Rat High Affinity Nerve Growth Factor Receptor (TrkA) siRNA
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Specifications
Specifications
| Accession Number | NP_067600 |
| Cell Lines Tested | Pc12 Rat Pheocromocytoma Cells from the Adrenal Medulla and Rat Primary Neurons. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | Ntrk1 |
| Gene ID (Entrez) | 59109 |
| Product Type | siRNA |
| Quantity | 10 nmol |
| Synonym | TrkA, High Affinity NGFR, NGFR, Trk |
| Target | Rat |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction