Learn More
enQuireBio™ TranslationBlocker Rat Apoptosis regulator Bcl-2 siRNA
Product Code. 16099161
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. 16099161
Brand: enQuireBio™ QX112nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of Apoptosis regulator Bcl-2 from Rat.
Specifications
Accession Number NP_058689
Cell Lines Tested Pc12 Pheochromocytoma Cells of the Rat Adrenal Gland (Medulla).
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol Bcl2
Gene ID (Entrez) 24224
Product Type siRNA
Quantity 2 nmol
Synonym Bcl-2, PPP1R50
Target Rat
Form Lyophilized