Learn More
enQuireBio™ TranslationBlocker Human TNF alpha siRNA
Product Code. 16019191
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. 16019191
Brand: enQuireBio™ QX222nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of TNF alpha from Human.
Specifications
Accession Number NP_000585
Cell Lines Tested Adherent LPS Treated Human PBMCs and Adherent dsRNA Treated Human PBMCs.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol TNF
Gene ID (Entrez) 7124
Product Type siRNA
Quantity 2 nmol
Synonym DIF, TNFA, TNFSF2, TNLG1F, TNF-alpha, Tumor necrosis factor alpha, Tumor necrosis factor
Target Human
Form Lyophilized