Learn More
enQuireBio™ TranslationBlocker Human TLR9 / CD289 siRNA
Product Code. 16039201
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. Quantity unitSize
16039201 2 nmol 2nmol
16029201 10 nmol 10nmol
2 options
Product Code. 16039201
Brand: enQuireBio™ QX282nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of TLR9 / CD289 from Human.
Specifications
Accession Number NP_059138
Cell Lines Tested 293T Human Embryonic Kidney Cells with Large T Antigen.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol TLR9
Gene ID (Entrez) 54106
Product Type siRNA
Quantity 2 nmol
Synonym CD289, Toll-like receptor 9, TLR-9
Target Human
Form Lyophilized