Learn More
enQuireBio™ TranslationBlocker Human TLR4 siRNA
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Specifications
Specifications
| Accession Number | NP_612564 |
| Cell Lines Tested | HepG2 Human Carcinoma Cells from Liver. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | TLR4 |
| Gene ID (Entrez) | 7099 |
| Product Type | siRNA |
| Quantity | 2 nmol |
| Synonym | TLR-4, Toll-like Receptor 4, CD284 |
| Target | Human |
| Form | Lyophilized |