Learn More
enQuireBio™ TranslationBlocker Human TLR3 siRNA
Product Code. 16019211
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. 16019211
Brand: enQuireBio™ QX312nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of TLR3 from Human.
Specifications
Accession Number NP_003256.1
Cell Lines Tested A549 Human Adenocarcinoma cells from Basal Alveolar Epithelium.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol TLR3
Gene ID (Entrez) 7098
Product Type siRNA
Quantity 2 nmol
Synonym TLR-3, Toll-like Receptor 3, CD283
Target Human
Form Lyophilized