missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human TLR3 siRNA
Specifications
Specifications
| Accession Number | NP_003256.1 |
| Cell Lines Tested | A549 Human Adenocarcinoma cells from Basal Alveolar Epithelium. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | TLR3 |
| Gene ID (Entrez) | 7098 |
| Product Type | siRNA |
| Quantity | 10 nmol |
| Synonym | TLR-3, Toll-like Receptor 3, CD283 |
| Target | Human |
| Form | Lyophilized |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?