missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human PD-L1 / CD274 siRNA
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Specifications
Specifications
| Accession Number | NP_054862 |
| Cell Lines Tested | Hep3b Human Hepatocellular Carinoma Cells, RKO Human Colon Cancer Cells, and Primary Dendritic Cells (DCs) isolated from PBMCs. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | CD274 |
| Gene ID (Entrez) | 29126 |
| Product Type | siRNA |
| Quantity | 2 nmol |
| Synonym | PDL1, CD274, Programmed Cell Death 1 Ligand 1, PDCD1L1, PDCD1 Ligand 1, B7H1, B7-H1, PD-1 L1, PD-1 Ligand 1 |
| Target | Human |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction