Learn More
enQuireBio™ TranslationBlocker Human IRF1 siRNA
Product Code. p-7155936
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. Quantity unitSize
16059211 2 nmol 2nmol
16049211 10 nmol 10nmol
2 options
Product Code. 16059211
Brand: enQuireBio™ QX332nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of IRF1 from Human.
Specifications
Accession Number NP_002189
Cell Lines Tested U-2 OS Huan Osteosarcoma Epithelial Cells from Bone.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol IRF1
Gene ID (Entrez) 3659
Product Type siRNA
Quantity 2 nmol
Synonym IRF-1, Interferon Regulatory Factor 1, MAR
Target Human
Form Lyophilized