Learn More
enQuireBio™ TranslationBlocker Human IRAK4 siRNA
Product Code. 16059201
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. Quantity unitSize
16059201 2 nmol 2nmol
16049201 10 nmol 10nmol
2 options
Product Code. 16059201
Brand: enQuireBio™ QX292nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of IRAK4 from Human.
Specifications
Accession Number NP_057207
Cell Lines Tested 293T Human Embryonic Kidney Cells with Large T Antigen.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol IRAK4
Gene ID (Entrez) 51135
Product Type siRNA
Quantity 2 nmol
Synonym Interleukin-1 receptor-associated kinase 4, IRAK-4
Target Human
Form Lyophilized