Learn More
enQuireBio™ TranslationBlocker Human Interleukin-10 / IL-10 siRNA
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Specifications
Tekniske data
| Accession Number | NP_000563 |
| Cell Lines Tested | Dendritic Cells derived from adherent human PBMCs. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | IL10 |
| Gene ID (Entrez) | 3586 |
| Product Type | siRNA |
| Quantity | 10 nmol |
| Synonym | IL10A, IL-10, TGIF, CSIF, GVHDS |
| Target | Human |
| Form | Lyophilized |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion