missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Human iNOS siRNA

Product Code. 16019241
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
This item is not returnable. View return policy

Product Code. 16019241

Brand: enQuireBio™ QX452nmol

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Blocks transcription and translation of iNOS from Human.

Specifications

Accession Number NP_000616
Cell Lines Tested Raji Human Lymphoblastoma Cells.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol NOS2
Gene ID (Entrez) 4843
Product Type siRNA
Quantity 2 nmol
Synonym HEP-NOS, NOS2A, NOS, INOS, Nitric oxide synthase, inducible
Target Human
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.