missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human FADD siRNA
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Specifications
Specifications
| Accession Number | NP_003815 |
| Cell Lines Tested | 293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | FADD |
| Gene ID (Entrez) | 8772 |
| Product Type | siRNA |
| Quantity | 10 nmol |
| Synonym | Fas Associated Via Death Domain, Fas-Associating Death Domain-Containing Protein, Growth-Inhibiting Gene 3 Protein, Fas (TNFRSF6)-Associated via Death Domain, Protein FADD, MORT1, FAS-Associated Death Domain Protein |
| Target | Human |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction