Learn More
enQuireBio™ TranslationBlocker Human Estrogen receptor siRNA
Product Code. 16039171
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. Quantity unitSize
16039171 2 nmol 2nmol
16029171 10 nmol 10nmol
2 options
Product Code. 16039171
Brand: enQuireBio™ QX142nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of Estrogen receptor from Human.
Specifications
Accession Number NP_000116
Cell Lines Tested MCF7 Human Adenocarcinoma Cells from Breast Cancer.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol ESR1
Gene ID (Entrez) 2099
Product Type siRNA
Quantity 2 nmol
Synonym ESRA, ESR, NR3A1, ER, ESTRR, Era
Target Human
Form Lyophilized