Learn More
enQuireBio™ TranslationBlocker Human Estrogen receptor beta siRNA
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Specifications
Specifications
| Accession Number | NP_034287 |
| Cell Lines Tested | MCF7 Human Adenocarcinoma Cells from Breast Cancer. |
| Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
| Gene Symbol | ESR2 |
| Gene ID (Entrez) | 13983 |
| Product Type | siRNA |
| Quantity | 2 nmol |
| Synonym | ESR-BETA, NR3A2, ESTRB, Erb, ESRB, ER-BETA, ERBeta |
| Target | Human |
| Form | Lyophilized |