Learn More
enQuireBio™ TranslationBlocker Human Estrogen receptor beta siRNA
Product Code. 16069171
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. Quantity unitSize
16069171 10 nmol 10nmol
16079171 2 nmol 2nmol
2 options
Product Code. 16069171
Brand: enQuireBio™ QX1610nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of Estrogen receptor beta from Human.
Specifications
Accession Number NP_034287
Cell Lines Tested MCF7 Human Adenocarcinoma Cells from Breast Cancer.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol ESR2
Gene ID (Entrez) 13983
Product Type siRNA
Quantity 10 nmol
Synonym ESR-BETA, NR3A2, ESTRB, Erb, ESRB, ER-BETA, ERBeta
Target Human
Form Lyophilized