Learn More
enQuireBio™ TranslationBlocker Human ErbB4 / Her4 siRNA
Product Code. 16099181
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. 16099181
Brand: enQuireBio™ QX212nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of ErbB4 / Her4 from Human.
Specifications
Accession Number NP_005226
Cell Lines Tested T-47d Human Carcinoma Cells from Breast.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol ERBB4
Gene ID (Entrez) 2066
Product Type siRNA
Quantity 2 nmol
Synonym p180erbB4, HER4, ALS19, Erb-b4, ErbB-4
Target Human
Form Lyophilized