missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Human Caspase-3 siRNA

Product Code. 16019231
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. Quantity unitSize
16019231 2 nmol 2nmol
16009231 10 nmol 10nmol
2 options
This item is not returnable. View return policy

Product Code. 16019231

Brand: enQuireBio™ QX402nmol

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Blocks transcription and translation of Caspase-3 from Human.

Specifications

Accession Number NP_004337
Cell Lines Tested 293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol CASP3
Gene ID (Entrez) 836
Product Type siRNA
Quantity 2 nmol
Synonym SCA-1, CPP32, CPP32B CASP3
Target Human
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.