Learn More
enQuireBio™ TranslationBlocker Human Caspase-2 siRNA
Product Code. 16079221
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
Product Code. 16079221
Brand: enQuireBio™ QX392nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of Caspase-2 from Human.
Specifications
Accession Number NP_116765
Cell Lines Tested U-2 OS Human Osteosarcoma Epithelial Cells from Bone.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol CASP2
Gene ID (Entrez) 835
Product Type siRNA
Quantity 2 nmol
Synonym CASP-2, ICH1, PPP1R57, NEDD-2, NEDD2, CASP2
Target Human
Form Lyophilized