Learn More
enQuireBio™ TranslationBlocker Human Caspase-1 siRNA
Product Code. 16059221
Click to view available options
Quantity:
1 nmol
5 nmol
Unit Size:
1nmol
5nmol
Product Code. 16059221
Brand: enQuireBio™ QX385nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of Caspase-1 from Human.
Specifications
Accession Number NP_001214
Cell Lines Tested Human Primary Keratinocytes.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol CASP1
Gene ID (Entrez) 834
Product Type siRNA
Quantity 5 nmol
Synonym Interleukin 1, Beta, Convertase, ICE, Caspase 1
Target Human
Form Lyophilized